Learn More
Medchemexpress LLC Triiodothyronine sulfate | 31135-55-4 | 98.8% | 1 MG
SDP
Catalog No. 5000340823
Encompass_Preferred
Triiodothyronine sulfate is the main metabolite of the thyroid hormone triiodothyronine (T3). Triiodothyronine is an active form of thyroid hormone that binds to β1 thyroid hormone receptor (TRβ1) and activates its activity.
  • Stimulates the proliferation of hepatocarcinoma cells in which TRβ1 is overexpressed.
  • Binds to human β1 thyroid hormone receptor (hTRβ1) and changes its conformation.
  • Promotes growth, induces differentiation, and regulates metabolic effects.
Catalog No. 50-003-40823 Supplier Medchemexpress LLC Supplier No. HY1269961MG
Triiodothyronine sulfate is the main metabolite of the thyroid hormone triiodothyronine (T3). Triiodothyronine is an active form of thyroid hormone that binds to β1 thyroid hormone receptor (TRβ1) and activates its activity.
  • Stimulates the proliferation of hepatocarcinoma cells in which TRβ1 is overexpressed.
  • Binds to human β1 thyroid hormone receptor (hTRβ1) and changes its conformation.
  • Promotes growth, induces differentiation, and regulates metabolic effects.