Learn More
Medchemexpress LLC Triiodothyronine sulfate | 31135-55-4 | 98.8% | 10 MG
SDP
Catalog No. 5000342822
Encompass_Preferred
Triiodothyronine sulfate is the main metabolite of the thyroid hormone triiodothyronine (T3). T3, an active form of thyroid hormone, activates β1 thyroid hormone receptor (TRβ1) by binding to it. This product promotes growth, induces differentiation, and regulates metabolic effects.
  • Identified as the main metabolite of thyroid hormone triiodothyronine (T3)
  • Activates β1 thyroid hormone receptor (TRβ1)
  • Stimulates proliferation of hepatocarcinoma cells where TRβ1 is overexpressed in vitro
  • Promotes growth
  • Induces differentiation
  • Regulates metabolic effects
  • For research use only
Catalog No. 50-003-42822 Supplier Medchemexpress LLC Supplier No. HY12699610MG
Triiodothyronine sulfate is the main metabolite of the thyroid hormone triiodothyronine (T3). T3, an active form of thyroid hormone, activates β1 thyroid hormone receptor (TRβ1) by binding to it. This product promotes growth, induces differentiation, and regulates metabolic effects.
  • Identified as the main metabolite of thyroid hormone triiodothyronine (T3)
  • Activates β1 thyroid hormone receptor (TRβ1)
  • Stimulates proliferation of hepatocarcinoma cells where TRβ1 is overexpressed in vitro
  • Promotes growth
  • Induces differentiation
  • Regulates metabolic effects
  • For research use only