Learn More
Eurofins Discoverx Triiodothyronine (T3), 10 mg. (92-1137)
Catalog No. 501999669
Encompass
Triiodothyronine (T3) is a thyroid hormone that is an active form of thyroxine, and an agonist of the thyroid hormone receptors THRα and THRβ. It affects nearly every physiological process in the body, and has a vital role in metabolism, heart and digestive function, muscle control, brain development, and bone maintenance. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9669 Supplier Eurofins Discoverx Supplier No. 921137
Triiodothyronine (T3) is a thyroid hormone that is an active form of thyroxine, and an agonist of the thyroid hormone receptors THRα and THRβ. It affects nearly every physiological process in the body, and has a vital role in metabolism, heart and digestive function, muscle control, brain development, and bone maintenance. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.