Learn More
Biomatik Corporation Triiodothyronine (T3) CLIA Kit
Supplier:  Biomatik Corporation EKU08621
Encompass_Preferred
Triiodothyronine (T3) CLIA Kit for detecting T3 in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 78.1-20,000pg/ml. Sensitivity: 25.9pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Target Synonyms: 3,3',5-Triiodo-L-Thyronine; Liothyronine; Cytomel; Tertroxin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7667