Learn More
Arbor Assays Triiodothyronine (T3) ELISA Kit (5 Plate)
SDP
Supplier:  Arbor Assays K056H5
Encompass_Preferred
Quantitatively measures T3 present in urine, extracted serum, plasma, fecal extracts, and tissue culture media samples. It measures total T3 in extracted serum, plasma and fecal extracts.
Catalog No. 50-227-8483