Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triisobutylaluminum Solution

Catalog No. 423793100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-379-3100ML 100 mL
42-379-3800ML 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 42-379-3100ML Supplier Sigma Aldrich Supplier No. 423793100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triisobutylaluminum solution (TIBA) (1.0 M in Hexane) can be used as a reagent in the preparation of triisobutylaluminium immobilized ++ gels (poly(organosilicon) networks), which can be further used as cocatalyst in ethylene polymerization.

Specifications

CAS 100-99-2
Density 0.695 g/mL at 25°C
Boiling Point 68°C to 69°C
Molecular Formula C12H27Al
Linear Formula [(CH3)2CHCH2]3Al
MDL Number MFCD00008929
Quantity 100 mL
Synonym TIBA solution
Molecular Weight (g/mol) 198.32 g/mol
Biological Activity Central Nervous System, Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.