Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triisopropanolamine

Catalog No. 2547461KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-474-61KG 1 kg
25-474-6100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-474-61KG Supplier Sigma Aldrich Supplier No. 2547461KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triisopropanolamine

Specifications

CAS 122-20-3
Melting Point 48°C to 52°C
Boiling Point 190°C (23 mmHg)
Molecular Formula C9H21NO3
Linear Formula [CH3CH(OH)CH2]3 N
Quantity 1 kg
MDL Number MFCD00004533
Synonym 1,1 ,1 -Nitrilotri(-2-propanol); Tris(2-hydroxypropyl)amine
Molecular Weight (g/mol) 191.27
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.