Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triisopropanolamine, 95%

Catalog No. AAA199660E
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA199660E 2500 g
AAA1996636 500 g
AAA199660C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA199660E Supplier Thermo Scientific Chemicals Supplier No. A199660E
Only null left
Add to Cart

CAS: 122-20-3 | C9H21NO3 | 191.271 g/mol

Major applications include water-based coating applications and agricultural products. Additional applications are antistat agents for polymers, corrosion inhibitor, electrodeposition/electrocoating, lubricants, paper, pigment dispersion, plastics, polyurethane additive, reaction intermediates, rubber curing, surfactants, mineral dispersion, and urethanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Major applications include water-based coating applications and agricultural products. Additional applications are antistat agents for polymers, corrosion inhibitor, electrodeposition/electrocoating, lubricants, paper, pigment dispersion, plastics, polyurethane additive, reaction intermediates, rubber curing, surfactants, mineral dispersion, and urethanes.

Solubility
Insoluble in water. Soluble ethanol, diethyl ether, chloroform (slightly ), and methanol (>500 g/100g).

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-20-3
Molecular Formula C9H21NO3
Molecular Weight (g/mol) 191.271
MDL Number MFCD00004533
InChI Key SLINHMUFWFWBMU-UHFFFAOYSA-N
Synonym triisopropanolamine, tipa, tri-2-propanolamine, tri-iso-propanolamine, tris 2-propanol amine, 2-propanol, 1,1',1-nitrilotris, tris 2-hydroxypropyl amine, 1,1',1-nitrilotripropan-2-ol, tris 2-hydroxy-1-propyl amine, caswell no. 891
PubChem CID 24730
IUPAC Name 1-[bis(2-hydroxypropyl)amino]propan-2-ol
SMILES CC(CN(CC(C)O)CC(C)O)O

Specifications

Melting Point 48°C to 52°C
Boiling Point 189°C to 191°C (23 mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Linear Formula (CH3CHOHCH2)3N
Quantity 2500 g
Beilstein 1071570
Sensitivity Hygroscopic
Solubility Information Insoluble in water. Soluble ethanol,diethyl ether,chloroform (slightly ),and methanol (>500g/100g).
Formula Weight 191.27
Percent Purity 95%
Chemical Name or Material Triisopropanolamine
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 204-528-4
RTECSNumber UB8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.