Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triisopropanolamine, 98%

Catalog No. AC245760100
Change view
Click to view available options
Quantity:
100 g
1 kg
10 kg
Packaging:
Glass bottle
Plastic Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC245760100 10 kg Plastic Can
AC245761000 100 g Glass bottle
AC245760010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC245760100 Supplier Thermo Scientific Chemicals Supplier No. 245760100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 122-20-3 | C9H21NO3 | 191.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-20-3
Molecular Formula C9H21NO3
Molecular Weight (g/mol) 191.27
MDL Number MFCD00004533
InChI Key SLINHMUFWFWBMU-UHFFFAOYSA-N
Synonym triisopropanolamine, tipa, tri-2-propanolamine, tri-iso-propanolamine, tris 2-propanol amine, 2-propanol, 1,1',1-nitrilotris, tris 2-hydroxypropyl amine, 1,1',1-nitrilotripropan-2-ol, tris 2-hydroxy-1-propyl amine, caswell no. 891
PubChem CID 24730
IUPAC Name 1-[bis(2-hydroxypropyl)amino]propan-2-ol
SMILES CC(CN(CC(C)O)CC(C)O)O

Specifications

Melting Point 48.0°C to 52.0°C
Color White to Yellow
Boiling Point 190.0°C (23.0 mmHg)
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic Can
Linear Formula N(CH2CH(OH)CH3)3
Quantity 10 kg
Beilstein 04,762
Fieser 17,371; 12,445; 13,46
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, diethyl ether
Viscosity 138 mPa.s (60°C)
Formula Weight 191.27
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Triisopropanolamine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
EINECSNumber 204-528-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.