Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triisopropanolamine (mixture of isomer) 98.0+%, TCI America™
SDP

Catalog No. N0104500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0104500G 500 g
N010425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0104500G Supplier TCI America Supplier No. N0104500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 122-20-3
Molecular Formula C9H21NO3
Molecular Weight (g/mol) 191.271
MDL Number MFCD00004533
InChI Key SLINHMUFWFWBMU-UHFFFAOYSA-N
Synonym triisopropanolamine, tipa, tri-2-propanolamine, tri-iso-propanolamine, tris 2-propanol amine, 2-propanol, 1,1',1-nitrilotris, tris 2-hydroxypropyl amine, 1,1',1-nitrilotripropan-2-ol, tris 2-hydroxy-1-propyl amine, caswell no. 891
PubChem CID 24730
IUPAC Name 1-[bis(2-hydroxypropyl)amino]propan-2-ol
SMILES CC(CN(CC(C)O)CC(C)O)O

Specifications

Color White
Boiling Point 305°C
Quantity 500 g
Formula Weight 191.27
Percent Purity ≥98.0% (T)
Grade Not Available
Physical Form Crystalline Lumps
Chemical Name or Material Triisopropanolamine (mixture of isomer)
EINECSNumber (2)-0310
RTECSNumber UB8750000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.