Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triisopropyl Borate

Catalog No. 1973352KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
80 g
750 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-733-52KG 2 kg
19-733-5750G 750 g
19-733-580G 80 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 19-733-52KG Supplier Sigma Aldrich Supplier No. 1973352KG
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trialkyl borate with good thermal properties.Reagent used to ortho-borylate 1-substituted naphthalenes for Pd-catalyzed coupling with aryl halides.

Specifications

CAS 5419-55-6
Density 0.815 g/mL at 25°C
Boiling Point 139°C to 141°C
Molecular Formula C9H21BO3
Refractive Index n20/D 1.376
Linear Formula [(CH3)2CHO]3B
MDL Number MFCD00008872
Quantity 2 kg
Synonym Boric acid triisopropyl ester; Boron isopropoxide; Triisopropoxyborane
Molecular Weight (g/mol) 188.07 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.