Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triisopropyl phosphite

Catalog No. T67806500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T67806500ML 500 mL
T67806100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T67806500ML Supplier Sigma Aldrich Supplier No. T67806500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Triisopropyl phosphite can be used as a reactant:
  • With ru-based indenylidene complexes to form 1st generation complexes for metathesis reactions
  • In perkow-type reaction to synthesize compounds containing polarized carbon-carbon double bonds
  • For the synthesis of phosphonohydrazines by reacting with arylamines and isoamyl nitrite.it can also be used as an alternative to triphenylphosphine in mitsunobu reaction to facilitate the isolation of products

Specifications

CAS 116-17-6
Density 0.844 g/mL (at 25°C (literature))
Boiling Point 63°C to 64°C (11 mmHg)
Molecular Formula C9H21O3P
Refractive Index n20/D 1.411 (literature)
Linear Formula [(CH3)2 CHO]3 P
MDL Number MFCD00008874
Quantity 500 mL
Molecular Weight (g/mol) 208.24
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.