Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triisopropylphosphine, 98%

Catalog No. AA4168409
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4168409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4168409 Supplier Thermo Scientific Chemicals Supplier No. 4168409
Add to Cart

CAS: 6476-36-4 | C9H21P | 160.241 g/mol

Triisopropylphosphine is used as a ligand in organometallic chemistry. It is also used to produce chlor(triisopropyl)phosphonium- dimesylamid at temperature of 40°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6476-36-4
Molecular Formula C9H21P
Molecular Weight (g/mol) 160.241
MDL Number MFCD00015029
InChI Key IGNTWNVBGLNYDV-UHFFFAOYSA-N
Synonym triisopropylphosphine, tri-i-propylphosphine, phosphine, triisopropyl, phosphine, tris 1-methylethyl, tri-isopropylphosphine, triisopropylphosphane, trisisopropylphosphine, triisopropyl phosphine, acmc-20ajdu, pubchem6458
PubChem CID 80969
IUPAC Name tri(propan-2-yl)phosphane
SMILES CC(C)P(C(C)C)C(C)C

Specifications

Density 0.839 g/mL
Boiling Point 176°C to 178°C
Flash Point -17°C (1°F)
Odor Pungent
Refractive Index 1.467
Quantity 10 g
UN Number UN2845
Sensitivity Air Sensitive
Solubility Information Soluble in alkanes.
Formula Weight 160.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Triisopropylphosphine
Show More Show Less
Hazard Category H250-H314
Hazard Statement GHS H Statement
H250-H252-H314-H318
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P222-P260-P264b-P280-P301+P330+P331-P302+P334-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 229-333-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.