Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triisopropylsilane

Catalog No. 23378150G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-378-150G 50 g
23-378-110G 10 g
23-378-1250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 23-378-150G Supplier Sigma Aldrich Supplier No. 23378150G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used in a highly diastereoelective preparation of anti-1,2-diols catalyzed by a Ni(O) N-heterocyclic carbene complex.

Specifications

CAS 6485-79-6
Density 0.773 g/mL at 25°C
Boiling Point 84°C to 86°C35 mmHg, lit.)
Molecular Formula C9H22Si
Refractive Index n20/D 1.434
Linear Formula [(CH3)2CH]3SiH
MDL Number MFCD00009657
Quantity 50 g
Molecular Weight (g/mol) 158.36 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.