Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Triisopropylsilane | 6485-79-6 | MFCD00009657 | 25g
SDP

Catalog No. 5000267839
Encompass_Preferred

Triisopropylsilane 98% is a strong reducing agent

Catalog No. 50-002-67839 Supplier Medchemexpress LLC Supplier No. HYY141025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triisopropylsilane 98% is a strong reducing agent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.