Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (Triisopropylsilyl)acetylene

Catalog No. 3600315G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-003-15G 5 g
36-003-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-003-15G Supplier Sigma Aldrich Supplier No. 3600315G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Asymmetric addition of (triisopropylsilyl)acetylene to α,α,α,α-unsaturated carbonyl compounds in the presence of a cobalt/duphos catalyst is reported
  • A sonogashira coupling reaction between 1-bromo-3-iodo-5-tertbutylbenzene and (triisopropylsilyl)acetylene is reported.(triisopropylsilyl)acetylene may be used as reagent for the rhodium-catalyzed asymmetric alkynylation of various α,α-unsaturated ketones
  • It may be used as reagent in the enantioselective synthesis of α-alkynylated nitroalkanes

Specifications

CAS 89343-06-6
Density 0.813 g/mL (at 25°C (literature))
Boiling Point 50°C to 52°C (0.6 mmHg)
Molecular Formula C11H22Si
Refractive Index n20/D 1.4527 (literature)
Linear Formula [(CH3)2 CH]3 SiC///CH
MDL Number MFCD00075452
Quantity 5 g
Synonym Ethynyltriisopropylsilane
Molecular Weight (g/mol) 182.38
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.