Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triisopropylsilyl Chloride

Catalog No. 241725250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-172-5250G 250 g
24-172-510G 10 g
24-172-550G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 24-172-5250G Supplier Sigma Aldrich Supplier No. 241725250G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Use of TIPS substituted alkynes in a preparation of silyl vinyl ketenes from chromium carbene complexes.

Specifications

CAS 13154-24-0
Density 0.901 g/mL at 25°C
Boiling Point 198°C (739 mmHg, lit.)
Molecular Formula C9H21ClSi
Refractive Index n20/D 1.452
Linear Formula [(CH3)2CH]3SiCl
MDL Number MFCD00009656
Quantity 250 g
Synonym Chlorotriisopropylsilane; TIPSCl; Triisopropylchlorosilane; Triisopropylsilyl chloride
Molecular Weight (g/mol) 192.8 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.