Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triisopropylsilyl Trifluoromethanesulfonate

Catalog No. 24846010G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-846-010G 10 g
24-846-050G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 24-846-010G Supplier Sigma Aldrich Supplier No. 24846010G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used to prepare silyloxy acetylenes fro mLithium acetylides; the silyloxy acetylenes were then employed in protic acid-promoted cyclizations of arenes, alkynes and olefins.

Specifications

CAS 80522-42-5
Density 1.14 g/mL at 25°C
Boiling Point 45°C to 46°C (0.03 mmHg)
Molecular Formula C10H21F3O3SSi
Linear Formula CF3SO3Si[CH(CH3)2]3
Refractive Index n20/D 1.415
MDL Number MFCD00009913
Quantity 10 g
Synonym TIPS triflate; Trifluoromethanesulfonic acid triisopropylsilyl ester; Triisopropylsilyl triflate
Molecular Weight (g/mol) 306.42 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.