Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific Triisopropylsilylacetylene, 89343-06-6, MFCD00075452, 5g

Supplier:  Matrix Scientific 0872045G

Encompass_Preferred

Molecular Formula C11H22Si, Purity 97+%, Molecular Weight 182.38, Boiling Point 50-53/0.6mm

Catalog No. 50-681-473


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.