Learn More
TARGETMOL CHEMICALS INC TRILACICLIB HYDROCHLORID 5MG
SDP
Catalog No. 502581369
Encompass_Preferred
Also available in 1 mg 10 mg 50 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. Trilaciclib hydrochloride (G1T28 hydrochloride) is an inhibitor of CDK4/6 (IC50s 1 nM and 4 nM for CDK4 and CDK6). purity: 99%
Catalog No. 50-258-1369 Supplier TARGETMOL CHEMICALS INC Supplier No. T132025MG
Also available in 1 mg 10 mg 50 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. Trilaciclib hydrochloride (G1T28 hydrochloride) is an inhibitor of CDK4/6 (IC50s 1 nM and 4 nM for CDK4 and CDK6). purity: 99%