Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trilaciclib hydrochloride | 1977495-97-8 | 519.47 g/mol | C24H32Cl2N8O | 25 MG
SDP

Catalog No. 5000248110
Encompass_Preferred

Trilaciclib hydrochloride is the hydrochloride salt of trilaciclib, a selective CDK4/6 inhibitor used as a research reagent to study cell-cycle regulation and to investigate protection against chemotherapy-induced myelosuppression. Supplied as a solid for laboratory use; chemical identity supported by safety documentation and public compound databases.

  • Potent CDK4/6 inhibitor for cell-cycle studies.
  • Applicable to research on chemotherapy-induced myelosuppression.
  • Supplied as a solid hydrochloride salt for laboratory use.
  • Molecular formula C24H32Cl2N8O; molecular weight 519.47 g/mol.
  • For research use only; not for clinical or diagnostic purposes.

Catalog No. 50-002-48110 Supplier Medchemexpress LLC Supplier No. HY101467A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trilaciclib hydrochloride is the hydrochloride salt of trilaciclib, a selective CDK4/6 inhibitor used as a research reagent to study cell-cycle regulation and to investigate protection against chemotherapy-induced myelosuppression. Supplied as a solid for laboratory use; chemical identity supported by safety documentation and public compound databases.

  • Potent CDK4/6 inhibitor for cell-cycle studies.
  • Applicable to research on chemotherapy-induced myelosuppression.
  • Supplied as a solid hydrochloride salt for laboratory use.
  • Molecular formula C24H32Cl2N8O; molecular weight 519.47 g/mol.
  • For research use only; not for clinical or diagnostic purposes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.