Learn More
Medchemexpress LLC Trilaciclib hydrochloride | 1977495-97-8 | 99.7% | 519.47 g/mol | C24H32Cl2N8O | 1 MG
SDP
Catalog No. 5000248806
Encompass_Preferred
Trilaciclib hydrochloride is the hydrochloride salt of trilaciclib, an orally active CDK4/6 inhibitor supplied as a research-grade analytical standard. It is provided as a light yellow to yellow solid for biochemical, cell-based, and preclinical studies, with documented activity against CDK4 and CDK6 and batch-specific characterization available.
  • Orally active CDK4/6 inhibitor for cell-cycle arrest studies.
  • High purity suitable for analytical and preclinical research.
  • Documented potency: CDK4 IC50 ≈ 1 nM; CDK6 IC50 ≈ 4 nM.
  • Solid formulation for ease of handling and storage.
  • Batch-specific datasheet and COA available for quality verification.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-48806 Supplier Medchemexpress LLC Supplier No. HY101467A1MG
Trilaciclib hydrochloride is the hydrochloride salt of trilaciclib, an orally active CDK4/6 inhibitor supplied as a research-grade analytical standard. It is provided as a light yellow to yellow solid for biochemical, cell-based, and preclinical studies, with documented activity against CDK4 and CDK6 and batch-specific characterization available.
  • Orally active CDK4/6 inhibitor for cell-cycle arrest studies.
  • High purity suitable for analytical and preclinical research.
  • Documented potency: CDK4 IC50 ≈ 1 nM; CDK6 IC50 ≈ 4 nM.
  • Solid formulation for ease of handling and storage.
  • Batch-specific datasheet and COA available for quality verification.
  • Intended for research use only; not for human or clinical use.