Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trilaciclib hydrochloride | 1977495-97-8 | 99.7% | 519.47 g/mol | C24H32Cl2N8O | 100 MG
SDP

Catalog No. 5000247955
Encompass_Preferred

Trilaciclib hydrochloride is the hydrochloride salt of trilaciclib, a potent cyclin-dependent kinase 4/6 (CDK4/6) inhibitor supplied as a light yellow to yellow solid for research use. It is commonly used in preclinical studies of cell-cycle regulation and to evaluate strategies for reducing chemotherapy-induced myelosuppression.

  • High purity (99.7%).
  • Molecular weight 519.47 g/mol.
  • Molecular formula C24H32Cl2N8O.
  • Light yellow to yellow solid appearance.
  • Intended for research and analytical applications.

Catalog No. 50-002-47955 Supplier Medchemexpress LLC Supplier No. HY101467A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trilaciclib hydrochloride is the hydrochloride salt of trilaciclib, a potent cyclin-dependent kinase 4/6 (CDK4/6) inhibitor supplied as a light yellow to yellow solid for research use. It is commonly used in preclinical studies of cell-cycle regulation and to evaluate strategies for reducing chemotherapy-induced myelosuppression.

  • High purity (99.7%).
  • Molecular weight 519.47 g/mol.
  • Molecular formula C24H32Cl2N8O.
  • Light yellow to yellow solid appearance.
  • Intended for research and analytical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.