Learn More
Medchemexpress LLC Trilaciclib hydrochloride | 1977495-97-8 | MFCD30747890 | >98.0% | 519.47 g/mol | C24H32Cl2N8O | 500 MG
SDP
Catalog No. 5000247956
Encompass_Preferred
Trilaciclib hydrochloride is the hydrochloride salt of trilaciclib (G1T28), a selective CDK4/6 inhibitor used as a research compound in cell-cycle and oncology studies. It is supplied as a solid analytical standard suitable for in vitro experiments and characterization. Reported molecular weight is 519.47 g/mol, chemical formula C24H32Cl2N8O, and vendor-reported purity is ≥98%.
  • Selective CDK4/6 inhibitor for cell-cycle research.
  • Supplied as a hydrochloride salt, stable solid for handling.
  • High purity suitable for analytical and in vitro use.
  • Molecular weight 519.47 g/mol and defined chemical formula.
  • Available in small research quantities; verify pack size with supplier before ordering.
Catalog No. 50-002-47956 Supplier Medchemexpress LLC Supplier No. HY101467A500MG
Trilaciclib hydrochloride is the hydrochloride salt of trilaciclib (G1T28), a selective CDK4/6 inhibitor used as a research compound in cell-cycle and oncology studies. It is supplied as a solid analytical standard suitable for in vitro experiments and characterization. Reported molecular weight is 519.47 g/mol, chemical formula C24H32Cl2N8O, and vendor-reported purity is ≥98%.
  • Selective CDK4/6 inhibitor for cell-cycle research.
  • Supplied as a hydrochloride salt, stable solid for handling.
  • High purity suitable for analytical and in vitro use.
  • Molecular weight 519.47 g/mol and defined chemical formula.
  • Available in small research quantities; verify pack size with supplier before ordering.