Learn More
Selleck Chemical LLC Trilaciclib
SDP
Supplier:  Selleck Chemical LLC S83895MG
Encompass_Preferred
Trilaciclib is a highly potent selective and reversible cyclin-dependent kinase 4 and 6 (CDK4/6) inhibitor Trilaciclib inhibits CDK4/cyclin D1 and CDK6/cyclin D3 with IC50 of 1 nM and 4 nM respectively
Catalog No. 50-217-3356