Learn More
Medchemexpress LLC Trilaurin | 538-24-9 | 99.78% | 500 MG
SDP
Catalog No. 5000338885
Encompass_Preferred
Trilaurin is a lipid that occurs as an endogenous metabolite, presenting as a white to off-white solid. Its chemical characteristics include a molecular weight of 639.00 and a molecular formula of C39H74O6. Research indicates that it possesses the ability to inhibit the formation of neoplasms (tumors) that are initiated by dimethylbenzanthracene (DMBA) and subsequently promoted by croton oil. This compound is designated for research use only.
  • Inhibits neoplasm formation initiated by dimethylbenzanthracene (DMBA)
  • Used in various compound screening libraries
  • Component of Metabolism/Protease Compound Library
  • Component of Natural Product Library
  • Component of Anti-Cancer Compound Library
  • Component of Human Endogenous Metabolite Compound Library
  • Component of Lipid Compound Library
Catalog No. 50-003-38885 Supplier Medchemexpress LLC Supplier No. HY126573500MG
Trilaurin is a lipid that occurs as an endogenous metabolite, presenting as a white to off-white solid. Its chemical characteristics include a molecular weight of 639.00 and a molecular formula of C39H74O6. Research indicates that it possesses the ability to inhibit the formation of neoplasms (tumors) that are initiated by dimethylbenzanthracene (DMBA) and subsequently promoted by croton oil. This compound is designated for research use only.
  • Inhibits neoplasm formation initiated by dimethylbenzanthracene (DMBA)
  • Used in various compound screening libraries
  • Component of Metabolism/Protease Compound Library
  • Component of Natural Product Library
  • Component of Anti-Cancer Compound Library
  • Component of Human Endogenous Metabolite Compound Library
  • Component of Lipid Compound Library