Learn More
Trilink Bio Technologies Inc N1-Methylpseudouridine-5'-Triphosphate, 5 umole
SDP
Catalog No. NC0750660
Encompass
N1-Methyl-Pseudouridine-5'-Triphosphate is a modified NTP for incorporation into messenger RNAs (mRNA) using T7 RNA Polymerase. Incorporation of N1-methylpseudouridine can reduce the immunogenicity of the resulting mRNA.
Catalog No. NC0750660 Supplier Trilink Bio Technologies Inc Supplier No. N10815
N1-Methyl-Pseudouridine-5'-Triphosphate is a modified NTP for incorporation into messenger RNAs (mRNA) using T7 RNA Polymerase. Incorporation of N1-methylpseudouridine can reduce the immunogenicity of the resulting mRNA.