Learn More
TARGETMOL CHEMICALS INC Trilinolein 500MG
SDP
Catalog No. 502877345
Encompass_Preferred
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Trilinolein (1,2,3-Trilinoleoylglycerol) is a triacylglycerol and has been reported to provide a number of beneficial effects including reducing thrombogenicity and arrhythmias and increasing erythrocyte deformability. It is also an antioxidant. Purity 99.4%
Catalog No. 50-287-7345 Supplier TARGETMOL CHEMICALS INC Supplier No. T5212500MG
Add to Cart
Add to Cart
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Trilinolein (1,2,3-Trilinoleoylglycerol) is a triacylglycerol and has been reported to provide a number of beneficial effects including reducing thrombogenicity and arrhythmias and increasing erythrocyte deformability. It is also an antioxidant. Purity 99.4%