Learn More
Medchemexpress LLC Trilinolein | 537-40-6 | 98.0% | 50 MG
SDP
Catalog No. 5000339405
Encompass_Preferred
Trilinolein is an orally active triglyceride that inhibits the PI3K/Akt, Ras/MEK/ERK signaling pathways, and MMP-2. It reduces oxidative stress, induces apoptosis, and inhibits cell migration. This compound is applicable in research for cardiovascular disease, cerebrovascular disease (e.g., cerebral ischemia), and non-small cell lung cancer studies.
  • Inhibits PI3K/Akt, Ras/MEK/ERK, and MMP-2 pathways.
  • Reduces oxidative stress and promotes apoptosis.
  • Inhibits cell migration.
  • Enhances endothelial cell viability and antioxidant activity.
  • Mitigates cerebral infarction and neurological deficits in ischemia-reperfusion models.
  • Decreases intimal hyperplasia in mouse models.
Catalog No. 50-003-39405 Supplier Medchemexpress LLC Supplier No. HY12839350MG
Trilinolein is an orally active triglyceride that inhibits the PI3K/Akt, Ras/MEK/ERK signaling pathways, and MMP-2. It reduces oxidative stress, induces apoptosis, and inhibits cell migration. This compound is applicable in research for cardiovascular disease, cerebrovascular disease (e.g., cerebral ischemia), and non-small cell lung cancer studies.
  • Inhibits PI3K/Akt, Ras/MEK/ERK, and MMP-2 pathways.
  • Reduces oxidative stress and promotes apoptosis.
  • Inhibits cell migration.
  • Enhances endothelial cell viability and antioxidant activity.
  • Mitigates cerebral infarction and neurological deficits in ischemia-reperfusion models.
  • Decreases intimal hyperplasia in mouse models.