Learn More
Selleck Chemical LLC Citric acid trilithium salt tetrahydrate S4519-100mg
SDP
Catalog No. 503111539
Encompass_Preferred
Citric acid trilithium salt tetrahydrate (Trilithium citrate tetrahydrate Lithium citrate tribasic tetrahydrate) is a pharmaceutical and construction material It is commonly used in HPLC gradient elution for quantification of amino acids
Catalog No. 50-311-1539 Supplier Selleck Chemical LLC Supplier No. S4519100MG
Citric acid trilithium salt tetrahydrate (Trilithium citrate tetrahydrate Lithium citrate tribasic tetrahydrate) is a pharmaceutical and construction material It is commonly used in HPLC gradient elution for quantification of amino acids