Learn More
Charles River Laboratories Endosafe Trillium Recombinant Cascade Reagent, 6 vials per pack, 32 tests per vial, Kinetic chromogenic detection, 0.001-100 EU/mL range, Lyophilized
Supplier:  Charles River Laboratories RCRV3032PK
Encompass
Endosafe Trillium Recombinant Cascade Reagent, 6 vials per pack, 32 tests per vial, Kinetic chromogenic detection, 0.001-100 EU/mL range, Lyophilized
Catalog No. NC3640924