Learn More
TARGETMOL CHEMICALS INC TRILOBATIN 5MG
SDP
Catalog No. 502577021
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Trilobatin (P-Phlorizin) has anti-oxidant effect can increase superoxide dismutase (SOD) activity.Trilobatin has anti-inflammatory effect it potentially inhibits the lipopolysaccharide (LPS)-induced inflammatory response by suppressing the NF-(kappa)B signaling pathway.Trilobatin shows a strong inhibitory activity against (alpha)-glucosidase and a moderate inhibitory activity against (alpha)-amylase for management of postprandial hyperglycemia with less side effect. purity: 97%
Catalog No. 50-257-7021 Supplier TARGETMOL CHEMICALS INC Supplier No. T2S07315MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Trilobatin (P-Phlorizin) has anti-oxidant effect can increase superoxide dismutase (SOD) activity.Trilobatin has anti-inflammatory effect it potentially inhibits the lipopolysaccharide (LPS)-induced inflammatory response by suppressing the NF-(kappa)B signaling pathway.Trilobatin shows a strong inhibitory activity against (alpha)-glucosidase and a moderate inhibitory activity against (alpha)-amylase for management of postprandial hyperglycemia with less side effect. purity: 97%