Learn More
MSC Vermont Gage, Class ZZ Plus Plug Gage, 1.3010 in. Diameter, 1 ea.
Catalog No. 5000514650
Encompass
This Class ZZ Plus plug gage is designed for precision measurement and inspection tasks. It features a specified diameter of 1.3010 in. and is suitable for use in industrial metrology applications requiring strict dimensional accuracy.
  • Diameter of 1.3010 in.
  • Class ZZ Plus tolerance standard
  • Manufactured in the United States
  • Handle sold separately
Catalog No. 50-005-14650 Supplier MSC Supplier No. 64759210
This Class ZZ Plus plug gage is designed for precision measurement and inspection tasks. It features a specified diameter of 1.3010 in. and is suitable for use in industrial metrology applications requiring strict dimensional accuracy.
  • Diameter of 1.3010 in.
  • Class ZZ Plus tolerance standard
  • Manufactured in the United States
  • Handle sold separately