Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 1.2903 in. Diameter, Tool Steel, 1 ea.
Catalog No. 5000513851
Encompass
This Class ZZ plug gage features a diameter of 1.2903 in. and is engineered for precise measurement and inspection applications. Constructed from high-quality tool steel, it offers reliable performance and durability in industrial environments.
  • Diameter of 1.2903 in.
  • Constructed from tool steel with a hardness of 60/65 Rc
  • Tolerance of .0002 in. (0.005 mm)
  • Finish of 10 microinch or better
  • Roundness tolerance of .0001 in. (0.0025 mm)
  • NIST traceable with certificate of accuracy included
  • Handle sold separately
Catalog No. 50-005-13851 Supplier MSC Supplier No. 64758147
This Class ZZ plug gage features a diameter of 1.2903 in. and is engineered for precise measurement and inspection applications. Constructed from high-quality tool steel, it offers reliable performance and durability in industrial environments.
  • Diameter of 1.2903 in.
  • Constructed from tool steel with a hardness of 60/65 Rc
  • Tolerance of .0002 in. (0.005 mm)
  • Finish of 10 microinch or better
  • Roundness tolerance of .0001 in. (0.0025 mm)
  • NIST traceable with certificate of accuracy included
  • Handle sold separately