Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 1.2891 in. Diameter, 2 in. Length, 1 ea.
Catalog No. 5000512218
Encompass
This Class ZZ plug gage is designed for precision inspection and measurement applications. It features a diameter of 1.2891 in. and is manufactured from durable tool steel with a hardness of 60 to 65 Rc.
  • Diameter of 1.2891 in.
  • Constructed from tool steel hardened to 60/65 Rc
  • Finish of 10 microinch or better
  • Roundness of 0.0001 in. (0.0025 mm)
  • Marked with size and direction of tolerance
  • Includes a certificate of accuracy
  • NIST traceable
  • Handle sold separately
Catalog No. 50-005-12218 Supplier MSC Supplier No. 64758022
This Class ZZ plug gage is designed for precision inspection and measurement applications. It features a diameter of 1.2891 in. and is manufactured from durable tool steel with a hardness of 60 to 65 Rc.
  • Diameter of 1.2891 in.
  • Constructed from tool steel hardened to 60/65 Rc
  • Finish of 10 microinch or better
  • Roundness of 0.0001 in. (0.0025 mm)
  • Marked with size and direction of tolerance
  • Includes a certificate of accuracy
  • NIST traceable
  • Handle sold separately