Learn More
CELL MARQUE CORPORATION TRILOGY 1L
SDP
Catalog No. NC9894315
Encompass
Trilogy (with EDTA), 1L, Applications: Immunohistochemistry, Stable indefinitely at room temperature, Biodegradable and Non-Toxic.
Catalog No. NC9894315 Supplier CELL MARQUE CORPORATION Supplier No. 920P09
Trilogy (with EDTA), 1L, Applications: Immunohistochemistry, Stable indefinitely at room temperature, Biodegradable and Non-Toxic.