Learn More
CELL MARQUE CORPORATION Trilogy™ 20X Concentrated, Non-Toxic, Biodegradable, 200 mL
SDP
Catalog No. NC9909830
Encompass
Cell Marque Trilogy™ is a 20X concentrated, non-toxic, biodegradable solution tailored for immunohistochemistry (IHC). This EDTA-based pretreatment simplifies tissue preparation by efficiently combining deparaffinization, rehydration, and antigen unmasking in one step, eliminating the need for xylene. Its near-neutral pH enhances unmasking while preserving tissue integrity. This approach fosters standardized procedures, promoting consistent IHC staining results.
Catalog No. NC9909830 Supplier CELL MARQUE CORPORATION Supplier No. 920P06
Cell Marque Trilogy™ is a 20X concentrated, non-toxic, biodegradable solution tailored for immunohistochemistry (IHC). This EDTA-based pretreatment simplifies tissue preparation by efficiently combining deparaffinization, rehydration, and antigen unmasking in one step, eliminating the need for xylene. Its near-neutral pH enhances unmasking while preserving tissue integrity. This approach fosters standardized procedures, promoting consistent IHC staining results.