Learn More
Selleck Chemical LLC Trilostane 10mg 13647-35-3 WIN 24540
SDP
Supplier:  Selleck Chemical LLC S140410MG
Encompass_Preferred
Trilostane (WIN 24540) is an inhibitor of 3?-hydroxysteroid dehydrogenase used in the treatment of Cushing¿s syndrome. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-767-1