Learn More
Bachem H-Lys-Lys-Lys-OH
SDP
Catalog No. 501946806
Encompass_Preferred
H-Lys-Lys-Lys-OH Acetate, 13184-14-0, C18H38N6O4, Mr 402.54, 250mg. Trilysine has found use in medicine as component of polymeric hydrogel adhesives for wound closure and other surgical applications. Such hydrogels form in situ and can also serve as depot for the slow release of drugs.
Catalog No. 50-194-6806 Supplier Bachem Supplier No. 4002495.0250
Add to Cart
Add to Cart
H-Lys-Lys-Lys-OH Acetate, 13184-14-0, C18H38N6O4, Mr 402.54, 250mg. Trilysine has found use in medicine as component of polymeric hydrogel adhesives for wound closure and other surgical applications. Such hydrogels form in situ and can also serve as depot for the slow release of drugs.