Learn More
Electron Microscopy Sciences Diamond Cutter for Leica EM Trim New, 6 mm
SDP
Catalog No. 501931088
Encompass_Preferred
These cutters are used exclusively in the Leica EM Trim. These blades allow for parallel edges of the complete block which is mandatory for successful sectioning.
Size: 6 mm
Catalog No. 50-193-1088 Supplier Electron Microscopy Sciences Supplier No. DEMT
These cutters are used exclusively in the Leica EM Trim. These blades allow for parallel edges of the complete block which is mandatory for successful sectioning.
Size: 6 mm