Learn More
Grainger Trim Marked Fire Finish White
Catalog No. 502766664
Encompass_Preferred
The Grainger 16X368 is a 16-inch diameter, 368-tooth circular saw blade with a 5/8-inch arbor hole and a 0.062-inch kerf. It is designed for cutting wood and wood products. The blade has triple chip grind teeth and is made from high-speed steel for durability and long life. This circular saw blade can be used with table saws, miter saws, radial arm saws and other woodworking tools that accept a 5/8-inch arbor. It provides smooth, fast and accurate cuts in plywood, lumber, shelving, furniture and other wood materials. The 16X368 saw blade from Grainger is ideal for professional contractors as well as avid DIYers.
Catalog No. 50-276-6664 Supplier Grainger Supplier No. 16X368
The Grainger 16X368 is a 16-inch diameter, 368-tooth circular saw blade with a 5/8-inch arbor hole and a 0.062-inch kerf. It is designed for cutting wood and wood products. The blade has triple chip grind teeth and is made from high-speed steel for durability and long life. This circular saw blade can be used with table saws, miter saws, radial arm saws and other woodworking tools that accept a 5/8-inch arbor. It provides smooth, fast and accurate cuts in plywood, lumber, shelving, furniture and other wood materials. The 16X368 saw blade from Grainger is ideal for professional contractors as well as avid DIYers.