Learn More
Aviva Systems Biology Corp TRIM10 Antibody - C-terminal region (ARP34424_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34424_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TRIM10. Alternate names: RNF9, HERF1, RFB30ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-55386