Learn More
Aviva Systems Biology Corp TRIM10 Antibody - C-terminal region (ARP57848_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57848_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TRIM10. Alternate names: HERF1, MGC141979, RFB30, RNF9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-62161