Learn More
Aviva Systems Biology Corp TRIM14 Antibody - middle region (ARP34737_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34737_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIM14. Alternate names: KIAA0129ApplicationWB. Species reactivity: Horse, Human, Mouse, Rat
Catalog No. 50-014-55538