Learn More
Aviva Systems Biology Corp TRIM16 Antibody - N-terminal region (ARP38799_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38799_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM16. Alternate names: EBBPApplicationWB. Species reactivity: Human
Catalog No. 50-014-56805