Learn More
Aviva Systems Biology Corp TRIM17 Antibody - C-terminal region (ARP34548_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34548_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM17. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TRIM17. Alternate names: RBCC, terf, RNF16ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55445