Learn More
Aviva Systems Biology Corp TRIM21 Antibody - N-terminal region (ARP38247_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38247_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM21. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM21. Alternate names: RNF81, RO52, SSA, SSA1ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rat
Catalog No. 50-014-56579