Learn More
Sino Biological Recombinant Human TRIM21 Protein (His Tag)
Supplier:  Sino Biological 18010H07B100
Encompass_Preferred
A DNA sequence encoding the human TRIM21 (NP_003132.2) (Met1-Tyr475) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-211-4767