Learn More
Aviva Systems Biology Corp TRIM22 Antibody - middle region (ARP38734_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38734_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM22. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIM22. Alternate names: GPSTAF50, RNF94, STAF50ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-56773