Learn More
Aviva Systems Biology Corp TRIM23 Antibody - N-terminal region (ARP31998_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31998_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM23. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM23. Alternate names: ARD1, ARFD1, RNF46ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54412